Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICE1
Entrez Gene ID
488059
Gene Name
interactor of little elongation complex ELL subunit 1
Gene Aliases
-
Chromosome Location
Chr.34: 8052211 - 8110779 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022414372.1 XP_022270080.1 - 6577 122
XM_545182.5 XP_545182.3 - 6622 122

View Full Product Details