Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCHFR
Entrez Gene ID
487498
Gene Name
GTP cyclohydrolase I feedback regulator
Gene Aliases
-
Chromosome Location
Chr.30: 7900691 - 7904330 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_544622.6 XP_544622.2 2 - 3 257 115

View Full Product Details