Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTFP1
Entrez Gene ID
486352
Gene Name
mitochondrial fission process 1
Gene Aliases
-
Chromosome Location
Chr.26: 23565438 - 23568919 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_543478.6 XP_543478.2 2 - 3 421 103

View Full Product Details