Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSPG4
Entrez Gene ID
487658
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
-
Chromosome Location
Chr.30: 38465486 - 38498365 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005638641.3 XP_005638698.1 8 - 9 5393 72

View Full Product Details