Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMB6
Entrez Gene ID
607466
Gene Name
proteasome 20S subunit beta 6
Gene Aliases
-
Chromosome Location
Chr.5: 31776286 - 31778789 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844148.5 XP_849241.1 5 - 6 718 60

View Full Product Details