Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CISD3
Entrez Gene ID
491036
Gene Name
CDGSH iron sulfur domain 3
Gene Aliases
-
Chromosome Location
Chr.9: 23501167 - 23504302 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624585.3 XP_005624642.1 - 121 118

View Full Product Details