Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD7
Entrez Gene ID
607675
Gene Name
CD7 molecule
Gene Aliases
-
Chromosome Location
Chr.9: 45826 - 48864 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844435.5 XP_849528.1 - 700 61

View Full Product Details