Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOMM6
Entrez Gene ID
609079
Gene Name
translocase of outer mitochondrial membrane 6
Gene Aliases
-
Chromosome Location
Chr.12: 10473592 - 10484374 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_846282.4 XP_851375.1 2 - 3 263 110

View Full Product Details