Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYL1
Entrez Gene ID
610908
Gene Name
LYL1 basic helix-loop-helix family member
Gene Aliases
-
Chromosome Location
Chr.20: 49109358 - 49113911 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022407375.1 XP_022263083.1 1 - 2 1454 74

View Full Product Details