Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB10
Entrez Gene ID
487014
Gene Name
zinc finger and BTB domain containing 10
Gene Aliases
-
Chromosome Location
Chr.29: 27750151 - 27781995 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014109251.1 XP_013964726.1 5 - 6 1602 127

View Full Product Details