Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACADVL
Entrez Gene ID
489463
Gene Name
acyl-CoA dehydrogenase very long chain
Gene Aliases
-
Chromosome Location
Chr.5: 32189191 - 32194255 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_546581.5 XP_546581.3 14 - 15 1434 113

View Full Product Details