Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFYVE21
Entrez Gene ID
480447
Gene Name
zinc finger FYVE-type containing 21
Gene Aliases
-
Chromosome Location
Chr.8: 71451968 - 71462244 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005623860.3 XP_005623917.1 3 - 3 236 141
XM_537565.5 XP_537565.2 3 - 3 305 141
XM_863591.5 XP_868684.1 3 - 3 305 141

View Full Product Details