Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAPTM4B
Entrez Gene ID
475042
Gene Name
lysosomal protein transmembrane 4 beta
Gene Aliases
-
Chromosome Location
Chr.13: 41134 - 127399 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005627808.3 XP_005627865.1 - 765 146

View Full Product Details