Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FECH
Entrez Gene ID
610360
Gene Name
ferrochelatase
Gene Aliases
-
Chromosome Location
Chr.1: 18352184 - 18384251 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022416725.1 XP_022272433.1 - 296 170
XM_847843.5 XP_852936.2 - 563 170

View Full Product Details