Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTPD2
Entrez Gene ID
491241
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
-
Chromosome Location
Chr.9: 48589408 - 48594852 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116914.2 XP_013972389.1 1 - 2 168 139
XM_022423531.1 XP_022279239.1 1 - 2 348 139
XM_548362.5 XP_548362.1 1 - 2 168 139

View Full Product Details