Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNL4A
Entrez Gene ID
476156
Gene Name
thioredoxin like 4A
Gene Aliases
-
Chromosome Location
Chr.1: 722223 - 735253 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005615276.3 XP_005615333.1 - 296 137
XM_022407962.1 XP_022263670.1 - 302 137
XM_533363.6 XP_533363.2 - 270 137

View Full Product Details