Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR5A2
Entrez Gene ID
490252
Gene Name
nuclear receptor subfamily 5 group A member 2
Gene Aliases
-
Chromosome Location
Chr.7: 2925915 - 3062118 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005622276.3 XP_005622333.2 - 1037 122
XM_547371.6 XP_547371.4 - 1233 122

View Full Product Details