Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL3
Entrez Gene ID
475404
Gene Name
methyltransferase like 3
Gene Aliases
-
Chromosome Location
Chr.15: 18528755 - 18546303 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_532627.6 XP_532627.2 2 - 3 473 103

View Full Product Details