Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EEFSEC
Entrez Gene ID
476514
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA specific
Gene Aliases
-
Chromosome Location
Chr.20: 2138475 - 2405420 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022406312.1 XP_022262020.1 - 838 145
XM_533720.6 XP_533720.2 - 835 145

View Full Product Details