Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THNSL1
Entrez Gene ID
477983
Gene Name
threonine synthase like 1
Gene Aliases
-
Chromosome Location
Chr.2: 8482955 - 8497217 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616989.3 XP_005617046.1 - 769 99
XM_005616991.3 XP_005617048.1 - 608 99
XM_005616992.3 XP_005617049.1 - 597 99
XM_535167.6 XP_535167.1 - 687 99

View Full Product Details