Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
DUS4L
Entrez Gene ID
483264
Gene Name
dihydrouridine synthase 4 like
Gene Aliases
-
Chromosome Location
Chr.18: 13023755 - 13040180 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014120782.2 XP_013976257.1 - 624 126
XM_022405202.1 XP_022260910.1 - 528 126
XM_022405203.1 XP_022260911.1 - 384 126

View Full Product Details