Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STEAP3
Entrez Gene ID
483885
Gene Name
STEAP3 metalloreductase
Gene Aliases
-
Chromosome Location
Chr.19: 30745910 - 30795310 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005631912.3 XP_005631969.1 3 - 3 858 105
XM_005631913.2 XP_005631970.1 3 - 3 700 105
XM_005631915.3 XP_005631972.1 4 - 4 1046 105
XM_022406064.1 XP_022261772.1 5 - 5 1196 105

View Full Product Details