Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX3
Entrez Gene ID
477494
Gene Name
T-box transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.26: 11694885 - 11709594 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014107740.2 XP_013963215.1 - 2328 108

View Full Product Details