Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUCKS1
Entrez Gene ID
478944
Gene Name
nuclear casein kinase and cyclin dependent kinase substrate 1
Gene Aliases
-
Chromosome Location
Chr.38: 2157109 - 2188543 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005640796.3 XP_005640853.2 6 - 7 759 112
XM_022415448.1 XP_022271156.1 6 - 7 760 112

View Full Product Details