Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX11G
Entrez Gene ID
484992
Gene Name
peroxisomal biogenesis factor 11 gamma
Gene Aliases
-
Chromosome Location
Chr.20: 52284674 - 52292922 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_542110.4 XP_542110.1 1 - 2 90 68

View Full Product Details