Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBLN2
Entrez Gene ID
483936
Gene Name
cerebellin 2 precursor
Gene Aliases
-
Chromosome Location
Chr.1: 6378153 - 6384559 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_541056.6 XP_541056.4 - 373 120

View Full Product Details