Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRB1
Entrez Gene ID
490260
Gene Name
crumbs cell polarity complex component 1
Gene Aliases
-
Chromosome Location
Chr.7: 5277394 - 5490542 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005622293.3 XP_005622350.1 - 3231 110

View Full Product Details