The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_014113154.2 | XP_013968629.1 | 9 - 9 | 1655 | 129 |
XM_014113155.2 | XP_013968630.1 | 9 - 9 | 1736 | 129 |