Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CASR
Entrez Gene ID
488007
Gene Name
calcium sensing receptor
Gene Aliases
-
Chromosome Location
Chr.33: 25340822 - 25432814 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081509.2 NP_001074978.1 6 - 6 2172 123
XM_014110141.2 XP_013965616.1 8 - 8 3331 123
XM_014110142.2 XP_013965617.1 9 - 9 2926 123
XM_014110143.2 XP_013965618.1 7 - 7 2561 123
XM_022413726.1 XP_022269434.1 7 - 7 2902 123
XM_022413727.1 XP_022269435.1 8 - 8 3049 123
XM_022413728.1 XP_022269436.1 9 - 9 2876 123
XM_022413729.1 XP_022269437.1 8 - 8 3323 123
GenBank mRNA DQ366290.1 - 6 - 6 2172 123

View Full Product Details