Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HOTAIR
- Entrez Gene ID
-
100686390
- Gene Name
- HOX transcript antisense RNA
- Gene Aliases
-
-
- Chromosome Location
-
Chr.27: 1296506 - 1300023 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_131937.1 |
|
1 - 2 |
184 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
homeobox C11 |
HOXC11 |
|
-6.8kb
|
Anti-sense |
Chr. - 1287326 - 1289577 on Build CanFam3.1 |
Downstream |
homeobox C12 |
HOXC12 |
|
6.7kb
|
Anti-sense |
Chr. - 1305219 - 1307350 on Build CanFam3.1 |
View Full Product Details