Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scn1b
Entrez Gene ID
100758608
Gene Name
sodium channel, voltage-gated, type I, beta subunit
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615396: 130907 - 137953 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512219.1 XP_003512267.1 2 - 3 185 69

View Full Product Details