Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgf11
Entrez Gene ID
100759104
Gene Name
fibroblast growth factor 11
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613643: 1855728 - 1858798 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497244.2 XP_003497292.1 3 - 4 429 73

View Full Product Details