Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taz
Entrez Gene ID
100758692
Gene Name
tafazzin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615411: 63013 - 69153 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512275.2 XP_003512323.1 4 - 5 369 67
XM_003512276.2 XP_003512324.1 4 - 5 369 67
XM_007652799.1 XP_007650989.1 4 - 5 369 67
XM_007652800.1 XP_007650990.1 4 - 5 369 67
XM_007652801.1 XP_007650991.1 4 - 5 369 67
GenBank mRNA JI892031.1 - 5 - 6 67

View Full Product Details