Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpr82
Entrez Gene ID
100765377
Gene Name
G protein-coupled receptor 82
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613590: 3601538 - 3602521 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495370.1 XP_003495418.1 1 - 1 822 91

View Full Product Details