Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Leprel1
Entrez Gene ID
100752370
Gene Name
leprecan-like 1
Gene Aliases
I79_000857
Chromosome Location
Chr.Un|NW_003613596: 2315641 - 2468114 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642431.1 XP_007640621.1 9 - 10 1151 59
GenBank mRNA JI873838.1 - 9 - 10 1043 59

View Full Product Details