Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Znf8
Entrez Gene ID
103163647
Gene Name
zinc finger protein 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614721: 212012 - 216401 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650115.1 XP_007648305.1 1 - 2 577 69

View Full Product Details