Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scgb3a1
Entrez Gene ID
100766899
Gene Name
secretoglobin, family 3A, member 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614145: 110485 - 112886 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505371.1 XP_003505419.1 1 - 2 51 73

View Full Product Details