Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab31
Entrez Gene ID
100757810
Gene Name
RAB31, member RAS oncogene family
Gene Aliases
I79_016329
Chromosome Location
Chr.Un|NW_003614535: 474914 - 557258 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508397.2 XP_003508445.1 1 - 2 67 64

View Full Product Details