Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btbd1
Entrez Gene ID
100770759
Gene Name
BTB (POZ) domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615236: 271031 - 294543 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511810.2 XP_003511858.2 3 - 4 555 75
GenBank mRNA JI869323.1 - 3 - 4 555 75
JP044018.1 - 3 - 4 555 75

View Full Product Details