Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnk7
Entrez Gene ID
100763826
Gene Name
potassium channel, subfamily K, member 7
Gene Aliases
I79_021817
Chromosome Location
Chr.Un|NW_003615656: 214101 - 216609 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512933.1 XP_003512981.1 2 - 3 735 104

View Full Product Details