Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gimap8
Entrez Gene ID
100773700
Gene Name
GTPase, IMAP family member 8
Gene Aliases
I79_007704
Chromosome Location
Chr.Un|NW_003613842: 1566966 - 1581588 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501330.1 XP_003501378.1 2 - 3 743 90

View Full Product Details