Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Evx1
Entrez Gene ID
100763300
Gene Name
even-skipped homeobox 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613870: 160659 - 162866 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501995.1 XP_003502043.1 1 - 2 423 77

View Full Product Details