Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sntg2
Entrez Gene ID
100752603
Gene Name
syntrophin, gamma 2
Gene Aliases
I79_007805
Chromosome Location
Chr.Un|NW_003613848: 638111 - 784345 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501414.2 XP_003501462.1 8 - 9 564 60

View Full Product Details