Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Znf3
Entrez Gene ID
100774228
Gene Name
zinc finger protein 3
Gene Aliases
I79_018694
Chromosome Location
Chr.Un|NW_003614875: 160312 - 171951 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510416.2 XP_003510464.1 2 - 3 361 82
XM_007651034.1 XP_007649224.1 2 - 3 611 82

View Full Product Details