Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Necap1
Entrez Gene ID
100774802
Gene Name
NECAP endocytosis associated 1
Gene Aliases
I79_018439
Chromosome Location
Chr.Un|NW_003614841: 212657 - 220281 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650811.1 XP_007649001.1 1 - 2 193 63
GenBank mRNA JI888107.1 - 1 - 2 193 63

View Full Product Details