Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lyl1
Entrez Gene ID
100757879
Gene Name
lymphoblastic leukemia associated hematopoiesis regulator 1
Gene Aliases
I79_008354
Chromosome Location
Chr.Un|NW_003613865: 533831 - 537811 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501900.2 XP_003501948.2 3 - 4 532 90

View Full Product Details