Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rccd1
Entrez Gene ID
100758931
Gene Name
RCC1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616071: 152544 - 157219 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653933.1 XP_007652123.1 2 - 3 523 65

View Full Product Details