Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam26d
Entrez Gene ID
100770005
Gene Name
family with sequence similarity 26, member D
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614863: 32423 - 35208 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510324.1 XP_003510372.1 1 - 2 568 85

View Full Product Details