Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmco2
Entrez Gene ID
100752424
Gene Name
transmembrane and coiled-coil domains 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613862: 113513 - 117248 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501815.2 XP_003501863.1 1 - 2 237 86

View Full Product Details