Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aif1l
Entrez Gene ID
100752261
Gene Name
allograft inflammatory factor 1-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615645: 42836 - 65682 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512897.1 XP_003512945.1 4 - 5 207 96

View Full Product Details